Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega KDR Kinase Enzyme System

Recombinant human KDR (amino acids 789-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. KDR (or kinase insert domain receptor) is a growth factor receptor tyrosine kinase originally isolated from human endothelial cells.

$540.50 - $1070.00

Specifications

Format Fluid
Molecular Weight 110
View More Specs

Includes: KDR Kinase Enzyme System (V2681)
  • Kinase: KDR, 10μg (Human, recombinant; amino acids 789–end), M.W.: ∽110kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT
ADP-Glo™ Kinase Assay and KDR Kinase Enzyme System (V9471)
  • 1 x 10μg KDR Kinase Enzyme System (V2681) and 1 x 1000 ADP-Glo™ Kinase Assays (V9471)

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV2681
View Documents
Promega
V2681
10 μg
Each for $540.50
Add to cart
 
PRV9471
View Documents
Promega
V9471
1/Ea.
Each for $1,070.00
Add to cart
 
Description

Description

  • Easily Screen and Profile KDR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

Fluid
110
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit