Learn More
Promega Magne™ HaloTag™ Magnetic Affinity Beads
The Magne HaloTag™ Beads provide a convenient method to covalently capture HaloTag™ fusion proteins with magnetic particles for protein pull-downs and purification. The beads offer high binding capacity with low nonspecific protein binding.
Supplier:  Promega G7282
 View more versions of this product
Catalog No. PRG7282
Add to cart
Description
Description
  • Magnetic Capture of HaloTag(R) Fusion Proteins for Pull-Downs and Purification
  • Bind ≥20mg of purified HaloTag(R) fusion protein per mL of particles
  • Manual or high-throughput applications
Specifications
Specifications
20% Slurry in 10 to 15% Ethanol
50 to 80 μm
Auto mated and manual purification of HaloTag™ fusion Proteins
2°C to 10°C
5 mL
Beads