Learn More
Promega MET Kinase Enzyme System
Recombinant human MET (amino acids 956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MET is a proto-oncogene that encodes a transmembrane growth factor receptor.
$540.50 - $1044.00
Includes: MET Kinase Enzyme System (V3361)
  • Kinase: MET, 10μg (Human, recombinant; amino acids 352–end), M.W.: ∽81kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT
ADP-Glo™ Kinase Assay and MET Kinase Enzyme System (V9571)
  • 1 x 10μg MET Kinase Enzyme System (V3361) and 1 x 1000 ADP-Glo™ Kinase Assays (V9571)
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3361
View Documents
Promega
V3361
10 μg
Each for $540.50
Add to cart
PRV9571
View Documents
Promega
V9571
1/Ea.
Each for $1,044.00
Add to cart
Description
Description
  • Easily Screen and Profile MET Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity