Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Minicell Adapter Kit for QuantiFluor™-ST Fluorometer
The QuantiFluor Fluorometers are small, affordable and sensitive instruments, providing easy and accurate fluorescence measurements. Both instruments provide high sensitivity for fluorescence measurements within minutes.
Manufacturer: Promega E6112
Description
- For Use With QuantiFluor(R)-ST and QuantiFluor(R)-P Instruments
- Instruments are discontinued
- Accessories available until March 1, 2019
Specifications
Store at 5 to 40°C | |
QuantiFluor Single-Tube Fluorometer | |
Measuring 50 to 250μL of sample |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title