Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega NAD(P)H-Glo™ Detection System
The NAD(P)H-Glo™ Detection System is a bioluminescent assay that measures the concentration of the reduced forms of nicotinamide adenine dinucleotides, NADH and NADPH, and does not discriminate between them.
$691.00 - $2897.00
Specifications
Content And Storage | -20°C |
---|---|
Detection Method | Bioluminescence |
pH | 7.5 |
Form | Fluid |
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Kit Contents | Quantity | Price | Quantity & Availability | ||||
PRG9061
|
Promega
G9061 |
1 x 55μL Reductase, 1 x 55μL reductase substrate, 1 x 1 Luciferin detection Reagent, 1 x 10mL Reconstitution Buffer | 10 mL |
Each for $691.00
|
|
||||
PRG9062
|
Promega
G9062 |
1 x 275μL Reductase, 1 x 275μL reductase substrate, 1 x 1 Luciferin detection Reagent, 1 x 50mL Reconstitution Buffer | 50 mL |
Each for $2,897.00
|
|
||||
Description
- Monitor the Concentration of Reduced Forms of NADH and NADPH
- Highly sensitive and rapid bioluminescent assay
- Easily adapted for high-throughput format
Specifications
-20°C | |
Bioluminescence | |
7.5 | |
Fluid |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title