Learn More
Promega Nicotinamide
The SIRT-Glo™ Assay is a single-reagent-addition, homogeneous, luminescent assay that measures the relative activity of the NAD+-dependent histone deacetylase (HDAC) class III enzymes (sirtuins; SIRTs) from purified enzyme sources.
Manufacturer:  Promega G6540
Catalog No. PRG6540
Add to cart
Description
Description
  • Detects Activity of NAD+-Dependent HDAC Class III Enzymes and SIRTs
  • Simple add-mix-measure protocol
  • Measure deacetylating activities from purified enzyme sources
Specifications
Specifications
10mL
8
−30 to −10°C
Fluid
1 x 30μL Nicotinamide 1M