Learn More
Promega PDGFRα Kinase Enzyme System
Recombinant human PDGFR alpha (amino acids 550-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR alpha is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.
$528.00 - $1070.00
Specifications
Storage Requirements <−65°C
View More Specs
Includes:
  • Kinase: PDGFRα, 10μg (Human, recombinant; amino acids 550–end), M.W.: ∽95kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT
Products 2
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Quantity Price Quantity & Availability  
PRV3721
View Documents Promotion Details
Promega
V3721
PDGFRα Kinase Enzyme System 10μg
Each for $528.00
Add to cart
PRV8011
View Documents Promotion Details
Promega
V8011
ADP-Glo™ Kinase Assay + PDGFRα Kinase Enzyme System 1/ea
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile PDGFRα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
<−65°C