Learn More
Promega PDGFRβ Kinase Enzyme System
Recombinant human PDGFR beta (amino acids 557-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR beta is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.
$540.50 - $1070.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3731
View Documents
Promega
V3731
10 μg
Each for $540.50
Add to cart
PRV8021
View Documents
Promega
V8021
Each
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile PDGFRβ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity