Learn More
Promega PDK1 Kinase Enzyme System
Recombinant full-length human PDK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. PDK1 (3-phosphoinositide-dependent protein kinase) is activated by the presence of PtdIns(3,4,5)P3 or PtdIns(3,4)P2.
$559.00 - $1153.00
Specifications
Product Type PDK1 Kinase Enzyme system
Includes 10μg Recombinant PDK1 Kinase, PDKtide (KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC) Substrate, Reaction Buffer, DTT and MnCl2
Molecular Weight 67
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Content And Storage Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Content And Storage Price Quantity & Availability  
PRV9681
View Documents
Promega
V9681
1/ea COMPONENTS AT DIFFERENT TEMPS
Each for $1,153.00
Add to cart
PRV2761
View Documents
Promega
V2761
10μg <−65°C
Each for $559.00
Add to cart
Description
Description
  • Easily Screen and Profile PDK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
PDK1 Kinase Enzyme system
10μg Recombinant PDK1 Kinase, PDKtide (KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC) Substrate, Reaction Buffer, DTT and MnCl2
67