Learn More
Promega pFN29K His6HaloTag™ T7 Flexi Vector for E. coli and Cell-Free Protein Expression
These vectors are used for inducible expression of polyhistidine-HaloTag™ fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter. Use the Flexi Cloning System vectors for C- or N-terminal fusions.
Supplier: Promega G8331
Description
- HaloTag(R)-Polyhistidine Dual Tag Bacterial Expression Vectors
- Express proteins in E. coli T7 cell-free expression systems
- Couple the protein solubility and labeling benefits of HaloTag(R) technology with the reusability and the throughput of Ni-affinity technology
- Select N- or C-terminal tags and either Ampicillin or Kanamycin resistance
Specifications
N-terminal His6-HaloTag™ dual tag | |
E. coli and T7 cell-free Protein expression systems | |
-30°C to -10°C |
pFC30A His-6HaloTag T7 Flexi Vector | |
20 μg | |
Bacterial Protein expression and purification, cell-free Protein expression and purification |