Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega PKCι Kinase Enzyme System

Recombinant full-length human PKC iota was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Protein kinase C iota is a member of the protein kinase C family of serine-threonine kinases.

$537.00 - $1062.00

Specifications

Format Fluid
Molecular Weight 98
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3751
View Documents
Promega
V3751
10 μg
Each for $537.00
Add to cart
 
PRV9751
View Documents
Promega
V9751
1/Ea.
Each for $1,062.00
Add to cart
 
Description

Description

  • Easily Screen and Profile PKCι Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications

Specifications

Fluid
98
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit