Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Proteasome-Glo™ Assays

The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in a purified enzyme preparation.

$1090.00

Specifications

Buffer Proteasome-Glo™ cell-based buffer
Quantity 50mL
Includes Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent
Storage Requirements −30 to −10°C
View More Specs

Includes: Buffer, substrate and luciferin detection reagent

Products 2
Catalog Number Mfr. No. Product Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type For Use With (Application) Price Quantity & Availability  
PRG8632
View Documents
Promega
G8632
Trypsin-like assay Measures Trypsin-like Protease activity associated with the Proteasome in a purified Enzyme-based format
Each for $1,090.00
Add to cart
 
PRG8642
View Documents
Promega
G8642
Caspase-like assay Measures Caspase-like Protease activity associated with the Proteasome in a purified Enzyme-based format
Each for $1,090.00
Add to cart
 
Description

Description

  • Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
  • Homogeneous, luminescent enzyme-based assays
  • Simple add-mix-measure protocol
  • Results in 10-30 minutes after reagent addition
Specifications

Specifications

Proteasome-Glo™ cell-based buffer
50mL
Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent
−30 to −10°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit