Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Proteasome-Glo™ Assays
Shop All Promega Corporation Products
Click to view available options
Product Type:
Caspase-like Assay
Trypsin-like Assay
Description
Includes
Buffer, substrate and luciferin detection reagent
- Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
- Homogeneous, luminescent enzyme-based assays
- Simple add-mix-measure protocol
- Results in 10-30 minutes after reagent addition
Specifications
Specifications
| Buffer | Proteasome-Glo™ cell-based buffer |
| Product Type | Caspase-like Assay |
| Quantity | 50 mL |
| Includes | Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent |
| Content And Storage | −30 to −10°C |
| For Use With (Application) | Measures Caspase-like Protease activity associated with the Proteasome in a purified Enzyme-based format |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction