Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega psiCHECK Vectors
psiCHECK-1 and psiCHECK-2 Vectors enable monitoring of changes in expression of a target gene fused to a reporter gene. They are used for optimizing RNA interference assays.
$498.50
Specifications
For Use With (Application) | Provide a quantitative and rapid approach for initial optimization of RNA interference (RNAi), monitors changes in expression of target gene |
---|---|
Quantity | 20μg |
Reporter Gene | Renilla Luciferase, Firefly Luciferase |
Storage Requirements | −30 to −10°C |
Catalog Number | Mfr. No. | Vector | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Vector | Price | Quantity & Availability | |||||
PRC8021
|
Promega
C8021 |
psiCHECK-2 vector |
Each for $498.50
|
|
Description
- Vectors for Optimizing RNA Interference Assays
- Monitor changes in expression of a target gene fused to a reporter gene; decreases in luciferase activity correlate with RNAi
- psiCHECK-1 Vector is used to monitor RNAi effects in live cells
- psiCHECK-2 Vector designed for endpoint lytic assays
Specifications
Provide a quantitative and rapid approach for initial optimization of RNA interference (RNAi), monitors changes in expression of target gene | |
20μg | |
Renilla Luciferase, Firefly Luciferase | |
−30 to −10°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title