Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega PureYield™ RNA Midiprep System

The PureYield RNA Midiprep System isolates intact, pure total RNA from essentially any sample type for use in a wide range of applications.

$225.00 - $2130.00

Specifications

Format Spin or Vacuum
For Use With (Application) Enables rapid purification of total RNA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRZ3370
View Documents
Promega
Z3370
1/Ea.
Each for $2,130.00
Add to cart
 
PRZ3740
View Documents
Promega
Z3740
10 Preps
Each for $225.00
Add to cart
 
Description

Description

  • High Yields of Total RNA Without Using Organic Solvents
  • Purifies total RNA directly from cultured cells, bacteria, blood, yeast, plants and other sample types
  • Undetectable genomic DNA contamination
  • Rapid, safe and efficient RNA purification method

U.S. Patent 6,194,562, Australian Patent 740145, Canadian Patent 2,329,067.

Specifications

Specifications

Spin or Vacuum
Enables rapid purification of total RNA
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit