Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega QuantiFluor™-ST and QuantiFluor-P Single-Tube Fluorometers: PCR Tube Adapter
The QuantiFluor Fluorometers are small, affordable and sensitive instruments, providing easy and accurate fluorescence measurements. Both instruments provide high sensitivity for fluorescence measurements within minutes.
Manufacturer: Promega E6101
Description
- For Use With QuantiFluor(R)-ST and QuantiFluor(R)-P Instruments
- Instruments are discontinued
- Accessories available until March 1, 2019
Certifications
CE marked
Specifications
Store at 5 to 40°C | |
1/ea | |
QuantiFluor-ST and QuantiFluor-P Single-Tube Fluorometers | |
PCR Tube Adapter |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title