Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Age I (K)

Catalog No. PRR7251
Click to view available options
Quantity:
100 U

Enzymes that recognize short, specific (often palindromic) DNA sequences in which the enzymes then cleave. (A/CCGG T T GGCC\A)

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix

Specifications

Concentration 3 to 10 U/μL
Components 1 x 1mL 10X Buffer K, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 100U AgeI, 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Capable of digesting DNA in <15 min.
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM NaCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 100 U
Cut Site A.CCGGT TGGCC.A
Product Type AgeI
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.