Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Bal I (G)

Specifications

Format Liquid
Concentration 2 to 10 U/μL
Incubator Temperature 37°C
pH 7.4
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
View More Specs

Products 1
Catalog Number Mfr. No. Components Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Components Quantity Price Quantity & Availability  
PRR6695
View Documents
Promega
R6695
1 x 1mL 10X Buffer G, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 250U BalI 250 U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Specifications

Specifications

Liquid
37°C
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
TGG.CCA ACC.GGT
2 to 10 U/μL
7.4
-20°C
BalI
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit