Learn More
Promega Restriction Enzymes - BamH I Blue/White Cloning Qualified (E)
Capable of digesting DNA in 15 minutes or less. Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. (G/GATC C C CTAG\G)
Supplier: Promega R4024
Description
- Active and capable of digestion directly in GoTaq™ Green Master Mix
- Ends generated with BamHI can be directly ligated to ends generated with BglII, BclI and XhoII; these restriction sites are not regenerated in the ligation product
Specifications
40 to 80 U/μL | |
37°C | |
Provides a higher level of quality control for Enzymes used in cloning applications | |
-20°C | |
G.GATCC CCTAG.G | |
Liquid |
1 x 1mL 10X Buffer E, 1 x 750μL Bovine Serum Albumin, Acetylated, 1 x 12500U BamHI (HC), 1 x 0.25mL Multi-Core™ 10X Buffer | |
7.4 | |
10mM Tris HCl (pH 7.4), 300mM KCl, 5mM MgCl2, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
12,500 U | |
BamHI (HC) |