Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzymes - Dde I (D)
Capable of digesting DNA in 15 minutes or less. DdeI produces an ambiguous extension, which is difficult to ligate. (C/TNA G G ANT\C)
$51.30 - $171.70
Specifications
For Use With (Application) | Capable of digesting DNA in <15 min. |
---|---|
Concentration | 10U/μL |
Cut Site | C.TNAG |
Format | Liquid |
Incubator Temperature | 37°C |
Catalog Number | Mfr. No. | Components | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Components | Quantity | Price | Quantity & Availability | ||||
PR-R6291
|
Promega
R6291 |
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 200U DdeI, 1 x 0.25mL Multi-Core™ 10X Buffer | 200U |
Each for $51.30
|
|
PR-R6295
|
Promega
R6295 |
Each for $171.70
|
|
Description
- Capable of digesting DNA in 15 minutes or less
Specifications
Capable of digesting DNA in <15 min. | |
C.TNAG | |
37°C | |
DdeI | |
−30 to −10°C |
10U/μL | |
Liquid | |
7.9 | |
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 0.01% Triton™ X-100, 50% Glycerol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title