Learn More
Promega Restriction Enzymes - Hinc II Blue/White Cloning Qualified (B)
Active and capable of digestion directly in GoTaq™ Green Master Mix. Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications. (GT(T/C)/(A/G)AC CA(A/G)\(T/C)TG)
Manufacturer:  Promega R6035
 View more versions of this product
Catalog No. PRR6035
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
Specifications
Specifications
HincII
10U/μL
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U HincII, 1 x 0.25mL Multi-Core™ 10X Buffer
1000U
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Liquid
GT(T/C).(A/G)AC CA(A/G).(T/C)TG
37°C
Provides a higher level of quality control for Enzymes used in cloning applications
−30 to −10°C