Learn More
Promega Restriction Enzymes - Hpa II (A)
HpaII and its isoschizomer, MspI, exhibit different methylation sensitivities. (C/CG G G GC\C)
Manufacturer:  Promega R6311
Catalog No. PR-R6311
Add to cart
Description
Description
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Specifications
HpaII (A)
10U/μL
1 x 1mL 10X Buffer A, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U HhaI, 1 x 0.25mL Multi-Core™ 10X Buffer
1000U
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
Liquid
C.CGG GGC.C
37°C
Exhibit different methylation sensitivities
−30 to −10°C