Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzymes - Kpn I Blue/White Cloning Qualified (J)
Capable of digesting DNA in 15 minutes or less. (G GTAC/C C\CATG G)
$102.20 - $212.50
Specifications
For Use With (Application) | Provides a higher level of quality control for Enzymes used in cloning applications |
---|---|
Concentration | 8 to 12U/μL |
Cut Site | GGTAC.C C.CATGG |
Format | Liquid |
Incubator Temperature | 37°C |
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity & Availability | ||||
PR-R6341
|
Promega
R6341 |
2,500U | 1 x 1mL 10X Buffer J, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2500U KpnI, 1 x 0.25mL Multi-Core™ 10X Buffer |
Each for $102.20
|
|
PR-R6345
|
Promega
R6345 |
Each for $212.50
|
|
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Provides a higher level of quality control for Enzymes used in cloning applications | |
GGTAC.C C.CATGG | |
37°C | |
KpnI | |
−30 to −10°C |
8 to 12U/μL | |
Liquid | |
7.3 | |
10mM Tris HCl (pH 7.4), 200mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title