Learn More
Promega Restriction Enzymes - Msp I (B)
MspI and its isoschizomer HpaII exhibit different methylation sensitivities.
Supplier: Promega R6401
Description
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Liquid | |
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 2000U MspI, 1 x 0.25mL Muti-Core™10X Buffer | |
7.3 | |
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
2000 U | |
MspI |
10 U/μL | |
37°C | |
MspI and its Isoschizomer HpaII exhibit different methylation sensitivities | |
-30°C to -10°C | |
C.CGG GGC.C |