Learn More
Promega Restriction Enzymes - Nco I Blue/White Cloning Qualified (D)
Capable of digesting DNA in 15 minutes or less. (C/CATG G G GTAC\C)
Supplier: Promega R6515
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Liquid | |
1 x 1mL 10X Buffer D, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U NcoI, 1 x 0.25mL Multi-Core™ 10X Buffer | |
7.4 | |
10mM Tris HCl (pH 7.4), 300mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
1000 U | |
NcoI |
10 U/μL | |
37°C | |
Provides a higher level of quality control for Enzymes used in cloning applications | |
-30°C to -10°C | |
C.CATGG GGTAC.C |