Learn More
Promega Restriction Enzymes - Nhe I Genome Qualified (B)
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications. (G/CTAG C C GATC\G)
Supplier: Promega R6501
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Genome qualified to ensure optimal performance in genomic analysis applications
†Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Liquid | |
1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 250U NheI, 1 x 0.25mL Multi-Core™ 10X Buffer | |
7.4 | |
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
250 U | |
NheI |
10 U/μL | |
37°C | |
Capable of digesting DNA in <15 min. | |
-30°C to -10°C | |
G.CTAGC CGATC.G |