Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzymes - Nhe I Genome Qualified (B)
Capable of digesting DNA in 15 minutes or less. Genome qualified to ensure optimal performance in genomic analysis applications. (G/CTAG C C GATC\G)
$106.80 - $252.50
Specifications
For Use With (Application) | Capable of digesting DNA in <15 min. |
---|---|
Concentration | 10U/μL |
Cut Site | G.CTAGC CGATC.G |
Format | Liquid |
Incubator Temperature | 37°C |
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Components | Price | Quantity & Availability | ||||
PR-R6501
|
Promega
R6501 |
250U | 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 250U NheI, 1 x 0.25mL Multi-Core™ 10X Buffer |
Each for $106.80
|
|
PRR6505
|
Promega
R6505 |
1,250U | 1 x 1mL 10X Buffer B, 1 x 1250μL Bovine Serum Albumin, Acetylated, 1 x 250U NheI, 1 x 0.25mL Multi-Core™ 10X Buffer |
Each for $252.50
|
|
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Genome qualified to ensure optimal performance in genomic analysis applications
Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.
Specifications
Capable of digesting DNA in <15 min. | |
G.CTAGC CGATC.G | |
37°C | |
NheI | |
−30 to −10°C |
10U/μL | |
Liquid | |
7.4 | |
10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title