Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Sph I Blue/White Cloning Qualified (K)

Capable of digesting DNA in 15 minutes or less. (G CATG/C C\GTAC G)

$162.00 - $518.00

Specifications

Format Liquid
Concentration 10 U/μL
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Components Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Components Price Quantity & Availability  
PRR6265
View Documents
Promega
R6265
1000 U 1 x 1mL 10X Buffer K, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U SphI
Each for $518.00
Add to cart
 
PR-R6261
View Documents
Promega
R6261
200 U 1 x 1mL 10X Buffer K, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 200U SphI
Each for $162.00
Add to cart
 
Description

Description

  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq(R) Green Master Mix
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Specifications

Liquid
37°C
Provides a higher level of quality control for Enzymes used in cloning applications
-30°C to -10°C
SphI
10 U/μL
7.4
10mM Tris HCl (pH 7.4), 100mM Sodium Citrate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
GCATG.C C.GTACG
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit