Learn More
Promega Restriction Enzymes - Sph I Blue/White Cloning Qualified (K)
Capable of digesting DNA in 15 minutes or less. (G CATG/C C\GTAC G)
Supplier: Promega R6261
This item is not returnable.
View return policy
Description
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq(R) Green Master Mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
Specifications
10 U/μL | |
37°C | |
Provides a higher level of quality control for Enzymes used in cloning applications | |
-30°C to -10°C | |
GCATG.C C.GTACG | |
Liquid |
1 x 1mL 10X Buffer K, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 200U SphI | |
7.4 | |
10mM Tris HCl (pH 7.4), 100mM Sodium Citrate, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol | |
200 U | |
SphI |