Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega Restriction Enzymes - Tru9 I (F)
Source: Thermus ruber 9
Supplier: Promega R7011
Includes: Tru9 I, 200U; Buffer F (10X), 1mL; MULTI-CORE™ Buffer (10X), 0.25mL; BSA, 150μL
Description
- Storage Buffer: 20mM Tris-HCl (pH 7.5), 50mM KCl, 0.1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% glycerol
Specifications
Liquid | |
1 x 1mL 10X Buffer F, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 200U Tru9I, 1 x 0.25mL Multi-Core™ 10X Buffer | |
7.5 | |
-20°C | |
T.CGA AGC.T |
8 to 12 U/μL | |
37°C | |
20mM Tris HCl (pH 7.5), 50mM KCl, 0.1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol | |
200 U | |
Taql |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title