Learn More
Promega Riboprobe™ Combination Systems
The Riboprobe Systems are designed to prepare high-specific-activity single-stranded RNA probes or microgram quantities of defined RNA transcripts from cloned DNA inserts.
$415.00
Specifications
For Use With (Application) Generation of high-specific-activity RNA probes, generation of microgram quantities of RNA transcripts, generation of RNA processing substrates, generation of antisense RNA
Quantity 1 System
Storage Requirements −30 to −10°C
View More Specs
Includes: RNA polymerase(s); pGEM™ Express Positive Control Template; RQ1 RNase-free DNase; Recombinant RNasin™ Ribonuclease Inhibitor; rATP; rCTP; rGTP; rUTP; DTT; Transcription-optimized buffer, 5X; Nuclease-free water
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
PR-P1450
View Documents
Promega
P1450
500U T3 RNA Polymerase, 500U T7 RNA Polymerase, 5μg pGEM™ express positive control template, 500μL transcription optimized 5X Buffer, 2500U recombinant RNasin™ R
Each for $415.00
Add to cart
PR-P1460
View Documents
Promega
P1460
500U SP6 RNA Polymerase, 500U T7 RNA Polymerase, 5μg pGEM™ express positive control template, 500μL transcription optimized 5X Buffer, 2500U recombinant RNasin™ R
Each for $415.00
Add to cart
Description
Description
  • Produce High-Specific-Activity RNA Probes or Defined RNA Transcripts
  • Choose from combination systems with T3/T7 or SP6/T7 RNA polymerases
  • SP6, T7 and T3 RNA Polymerases are extremely promoter-specific, allowing production of virtually homogeneous RNA using plasmid DNA as a template
  • Allows synthesis of RNA corresponding to the coding or noncoding strand of cloned DNA from a single plasmid construct
U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
Australian Patents 616881 and 641261 and other pending and issued patents, which claim vectors encoding a portion of human placental ribonuclease inhibitor, are exclusively licensed to Promega Corporation.
Specifications
Specifications
Generation of high-specific-activity RNA probes, generation of microgram quantities of RNA transcripts, generation of RNA processing substrates, generation of antisense RNA
1 System
−30 to −10°C