Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega SAM2 Biotin Capture Membrane
Rapidly bind biotinylated molecules based on their affinity for streptavidin
$413.71
Specifications
For Use With (Application) | Protein kinase activity quantitation, specific mRNA quantitation, cytokine quantitation, and specific gene quantitation |
---|
Catalog Number | Mfr. No. | Length (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Height (Metric) | Price | Quantity & Availability | ||||
PR-V2861
|
Promega
V2861 |
10.5 cm | 15 cm |
Each for $413.71
|
|
Description
- Provide quantitative, linear substrate binding in the nmol/cm2 range due to the high density of streptavidin on the membrane, permitting kinetic studies
- Optimized for low nonspecific binding
- Can be used to analyze a wide variety of biotinylated substances without optimizing each substrate for binding to a matrix
- Bind within one minute
- Provide linear binding of substrates up to the nmol/cm2 range, permitting kinetic studies
- Ideal for protein kinase activity quantitation, specific mRNA quantitation, cytokine quantitation, and specific gene quantitation
SAM2 Biotin Capture Membrane
- Available as a prenumbered, partially cut sheet, which permits easy separation into 96 individual squares
- Also available as an uncut sheet, which allows for sample application using a multichannel pipetter
- Can be analyzed using phosphorimaging analysis, autoradiography, scintillation counting, or chemiluminescent detection to quantitate results
- Contains SAM2 Biotin Capture Membrane in the wells of a microfiltration plate
- Allows washes to be performed using a vacuum manifold or a plate washer
- Supplied with a transparent top seal and an opaque bottom seal, allowing addition of scintillation fluid for data quantitation using a microplate liquid scintillation counter
Specifications
Protein kinase activity quantitation, specific mRNA quantitation, cytokine quantitation, and specific gene quantitation |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title