Learn More
Promega Sephacryl™ S-400
Reagent used for the synthesis of double-stranded cDNA from mRNA and subsequent ligation into a suitable vector.
Supplier: Promega V3181
Description
- Quality tested and certified free of DNase and RNase activity
Specifications
2°C to 10°C | |
Suspension in 10mM Tris-HCl (pH 8.0), 100mM NaCl and 1mM EDTA. | |
10 mL |
Liquid | |
For removal of excess adaptors for ligated products |