Learn More
Promega SYK Kinase Enzyme System
Recombinant full-length human SYK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SYK is a non-receptor protein tyrosine kinase that is widely expressed in hematopoietic cells.
$541.00 - $1070.00
Specifications
Molecular Weight 100
View More Specs
Includes: Kinase: Recombinant Human SYK, 10μg (full length), MW: ∽100kDa
Substrate: Poly (4:1 Glu, Tyr) Peptide
Other: Reaction Buffer, DTT
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3801
View Documents
Promega
V3801
10 μg
Each for $541.00
Add to cart
PRV8271
View Documents
Promega
V8271
1/Ea.
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile SYK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
100