Learn More
Promega TNT™ Coupled Reticulocyte Lysate Systems
TNT Coupled Reticulocyte Lysate Systems are single-tube, coupled transcription/translation systems for eukaryotic cell-free protein expression.
Supplier: Promega L4601
Includes: TNT Rabbit Reticulocyte Lysate, Appropriate TNT polymerase, Control DNA, TNT Reaction Buffer, Luciferase assay wells, Luciferase assay reagent, Amino acid mixtures
Description
- A Single-Tube, Coupled Transcription/Translation System
- Study protein:protein, protein:nucleic acid and protein expression interactions
- Generate proteins in one hour versus days
- You choose the promoter
Specifications
<-65°C | |
200μL TNT™ Rabbit Reticulocyte Lysate, 20μL TNT™ Reaction Buffer, 20μL TNT™ T7 or SP6 RNA Polymerase, 5μg Luciferase T7 or SP6 co | |
TnT SP6 coupled reticulocyte lysate system |
GST pull-downs, Gel shift assays, co-immunoprecipitation, characterization of Protein modifications, Protein-RNA interactions, Protein activity studies, confirmation of gene products, Protein truncation test (PTT) | |
8 Reactions - Trial Size |