Learn More
Promega ViviRen™ Live Cell Substrate
ViviRen Live Cell Substrate is a uniquely engineered coelenterazine that generates three- to fivefold brighter Renilla luciferase luminescence for super-sensitive quantitation of reporter gene, BRET and RNAi activity.
$224.00 - $10540.00
Specifications
Product Type Live Cell Substrate
View More Specs
Products 3
Catalog Number Mfr. No. Format Quantity Content And Storage Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Content And Storage Price Quantity & Availability  
PRE6491
View Documents
Promega
E6491
Liquid 0.37 mg <-65°C
Each for $224.00
Add to cart
PRE6492
View Documents
Promega
E6492
Solid 3.7 mg <-10°C
Each for $1,415.00
Add to cart
PRE6495
View Documents
Promega
E6495
Solid 37 mg <-10°C
Each for $10,540.00
Add to cart
Description
Description
  • Sensitive Quantitation of Renilla Luminescence in Live Cells
  • Engineered coelenterazine for live-cell reporter gene, BRET and RNAi assays
  • Permits real-time measurement in multiwell plates
  • High signal-to-background ratios
U.S. Pat. Nos. 7,268,229 and 7,537,912, Canadian Pat. No. 2,462,506 and other patents and patents pending.
Certain applications of this product may require licenses from others.
This product does not convey a license to use recombinant Renilla luciferase under U.S. Patent 5,418,155 and related patents. Promega offers licensed Renilla luciferase vectors, which may be used in conjunction with this product.
Specifications
Specifications
Live Cell Substrate