Learn More
Promega ZAP70 Kinase Enzyme System
Recombinant full-length human ZAP70 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ZAP70 is a nonreceptor protein tyrosine kinase (part of the Syk/Zap70 family) that is involved in signaling by the T-cell antigen receptor.
$540.50 - $1070.00
Specifications
Molecular Weight 96
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRV3811
View Documents
Promega
V3811
10 μg
Each for $540.50
Add to cart
PRV8311
View Documents
Promega
V8311
1000 Assays
Each for $1,070.00
Add to cart
Description
Description
  • Easily Screen and Profile ZAP70 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Specifications
96