Learn More
Corning™ Pronto!™ Plus Microarray Systems by CoorsTek™ and Promega™
Incorporate all of the components needed to perform a microarray experiment from RNA isolation to post-hybridization washing
Supplier: Corning™ 40055
Description
Convenient systems combine Promega SV Total RNA Isolation System, Promega ChipShot™ systems for direct-labeling and cleanup of cDNA (fluorescent dyes not included), and Corning Pronto! Microarray Printing and Hybridization Reagents. Deliver reliable and sensitive microarray results when used with DNA microarrays printed on UltraGAPS™ Slides.
• Optimized for 5μg of total RNA per labeling reaction
• Detect as little as 3pg target RNA
• Produce interslide CV less than 10%
• Utilize easy-to-follow protocols
Specifications
System 5 |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.