Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Marvel™ Pronto™ Single Pedestal Credenza

Ideal for office, classroom or administration areas

Manufacturer:  Marvel™ PCR6024SP/UT:OKPU

 View more versions of this product

Catalog No. 08-101-147



Description

Description

  • Melamine laminate top with 2 - 2 in. grommets
  • Full legs include 2 - 2 in. knockouts
  • Modesty panel with wire management locks with 2 keys, 1 hanging pedestal with box/file
  • Textured powder coat finish20 Gauge steel bases

Certifications

Greenguard, ISO 9001-2008 and UL Certified

Warranty and Services

Lifetime

Specifications

Specifications

Greenguard Certified; ISO 9001-2008 UL CERTIFIED
Putty
29 in.
Single Pedestal Desk
Oak
24 in.
Office, classroom or administration areas
20 gauge steel
60 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit