Learn More
Thermo Scientific™ ProPac™ IMAC-10 HPLC Columns
Resolve target proteins using a single column in a high-resolution gradient run with ProPac IMAC-10 LC Columns.
$1186.00
Specifications
Max. Pressure | 3000 psi (207 bar) |
---|---|
Particle Size | 10 μm |
pH | 2 to 12 |
Pore Size | Nonporous |
Temperature | 60°C |
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Column Format | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Column Format | Price | Quantity & Availability | ||
DX063276
|
Thermo Scientific™
063276 |
10 μm | 4 mm | 50 mm | Analytical Column |
Each for $1,186.00
|
|
||
DX063278
|
Thermo Scientific™
063278 |
10 μm | 4 mm | 250 mm | Analytical Column | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
DX063282
|
Thermo Scientific™
063282 |
10 μm | 22 mm | 250 mm | Loading Column | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
DX063718
|
Thermo Scientific™
063718 |
10 μm | 9 mm | 250 mm | Loading Column | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Description
Resolve target proteins using a single column in a high-resolution gradient run with Thermo Scientific™ ProPac™ IMAC-10 LC Columns. These analytical and semi-preparative HPLC columns provides high-purity separations of metal-binding proteins and high loading capacity for protein purification applications. The use of small, nonporous resin particles results in narrow, high-efficiency peaks, with minimal band broadening due to dispersion. Use loading columns to charge the ProPac IMAC-10 LC columns but avoid contact of the metal solution with the system.
Tailored Specificity for IMAC Separation of Proteins- Retention control by imidazole concentration or pH gradient
- Stable between pH 2 to 12, compatible with common protein purification reagents (nonionic detergents, reducing agents)
- Effective under native or denaturing HPLC conditions
- Wide range of metal-specific applications, including the separation of His-tagged protein aggregation variants, prion peptide analysis and separation of monoclonal antibodies
- Shipped free from any metal and ready to be charged with the metal of your choice.
- Reusable columns with metal tailored specificity
- Resolve target proteins using a single column in a high resolution gradient run
- High purity separations of metal-binding proteins
Specifications
3000 psi (207 bar) | |
2 to 12 | |
60°C | |
Spherical, Nonporous Polymer | |
Affinity Separation |
10 μm | |
Nonporous | |
Lysozyne Poly(IDA) Grafts | |
Separation of Metal-Binding Proteins |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.