Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propantheline Bromide, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
50-34-0
C23H30BrNO3
448.40
XLBIBBZXLMYSFF-UHFFFAOYSA-M
methylbis(propan-2-yl)[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium bromide
[Br-].CC(C)[N+](C)(CCOC(=O)C1C2=CC=CC=C2OC2=CC=CC=C12)C(C)C
50-34-0
C23H30BrNO3
448.40
XLBIBBZXLMYSFF-UHFFFAOYSA-M
methylbis(propan-2-yl)[2-(9H-xanthene-9-carbonyloxy)ethyl]azanium bromide
[Br-].CC(C)[N+](C)(CCOC(=O)C1C2=CC=CC=C2OC2=CC=CC=C12)C(C)C

P1509, 50-34-0, C23H30BrNO3

Spectrum™ Propantheline Bromide, USP is an antimuscarinic agent used for the treatment of excessive sweating (hyperhidrosis), cramps or spasms of the stomach, intestines (gut) or bladder, and involuntary urination. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 0.005
Quantity 25 g
Ignition Residue 0.001
CAS Max % 1
Percent Purity 98 to 102%
Grade USP