Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Proparacaine Hydrochloride 98.0+%, TCI America™
Click to view available options
Quantity:
1g
5g
Specifications
Quantity | 1g |
Melting Point | 184°C |
Color | White-Yellow |
Formula Weight | 330.85 |
Physical Form | Crystal-Powder at 20°C |
Percent Purity | ≥98.0% (HPLC,T) |
Chemical Name or Material | Proparacaine Hydrochloride |
Safety and Handling
RTECSNumber : DG3065000
TSCA : No