Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Propargyl benzoate, 98%

Manufacturer:  Alfa Aesar™ H5344414

Quantity 25g

 View more versions of this product

Catalog No. AAH5344414


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

6750-04-5
C6H5CO2CH2C≡CH
MFCD00996341
Propargyl benzoate
C10H8O2
160.17
Benzoic acid propargyl ester

Specifications

6750-04-5
C6H5CO2CH2C≡CH
MFCD00996341
225° to 226°C
Propargyl benzoate
C10H8O2
160.17
Benzoic acid propargyl ester
25g
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit