Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargyl bromide, 80 wt.% solution in toluene, stabilized, ACROS Organics™

Click to view available options
Quantity:
50mL
250mL
1L
Packaging:
AcroSeal™ Glass bottle
Glass bottle
106-96-7,108-88-3
C3H3Br
118.961
YORCIIVHUBAYBQ-UHFFFAOYSA-N
propargyl bromide,3-bromopropyne,3-bromo-1-propyne,1-propyne, 3-bromo,2-propynyl bromide,propynyl bromide,1-bromo-2-propyne,propyne, 3-bromo,gamma-bromoallylene,1-brom-2-propin
7842
3-bromoprop-1-yne
C#CCBr
106-96-7,108-88-3
C3H3Br
118.961
YORCIIVHUBAYBQ-UHFFFAOYSA-N
propargyl bromide,3-bromopropyne,3-bromo-1-propyne,1-propyne, 3-bromo,2-propynyl bromide,propynyl bromide,1-bromo-2-propyne,propyne, 3-bromo,gamma-bromoallylene,1-brom-2-propin
7842
3-bromoprop-1-yne
C#CCBr
Alkylation reagent
Linear Formula HC≡CCH2Br
Fieser 06,493; 10,332; 06,495
Density 1.3800g/mL
Solubility Information Solubility in water: immiscible.
Formula Weight 118.96
Physical Form Powder
Percent Purity 73 to 87% (propargyl bromide) (GC) 13 to 27% (toluene) (GC)
Packaging Glass bottle
Refractive Index 1.4900 to 1.4960
Solubility Solubility in water: immiscible.
Stabilizer 0.3% (MgO)
Specific Gravity 1.38
Boiling Point 88.0°C to 90.0°C
Color White
Quantity 50mL
Flash Point 4°C
Chemical Name or Material Propargyl bromide
Show More Show Less