Learn More
Propargyl ether, 98%, Thermo Scientific Chemicals
Description
Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1?-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Propargyl ether is a useful ether for proteomics research. It was used for the preparation of polyferrocenes, via polyaddition with 1,1′-bis(azidoethyl)ferrocene. It may be used for the preaparation of cyclic polystyrene.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Specifications
Density | 0.914 |
Boiling Point | 55°C to 57°C (65 mmHg) |
Flash Point | 24°C (75°F) |
Refractive Index | 1.443 |
Linear Formula | (HC≡CCH2)2O |
Quantity | 5 g |
UN Number | UN3271 |
Formula Weight | 94.11 |
Percent Purity | 98% |
Chemical Name or Material | Propargyl ether |
Safety and Handling
Hazard Category | H226-H302 |
Hazard Statement | GHS H Statement H301-H226 Toxic if swallowed. Flammable liquid and vapor. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P312-P303+P361+P353-P330-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHERS, N.O.S. |
RTECSNumber | UK5950000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.