Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propargylaldehyde diethyl acetal, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
10160-87-9
C7H12O2
128.17
MFCD00009237
RGUXEWWHSQGVRZ-UHFFFAOYSA-N
3,3-diethoxy-1-propyne,propargylaldehyde diethyl acetal,3,3-diethoxypropyne,propiolaldehyde diethyl acetal,1-propyne, 3,3-diethoxy,3,3-diethoxy-propyne,propynal diethyl acetal,1,1-diethoxyprop-2-yne,1-propyne,3,3-diethoxy,propargyl aldehyde diethylacetal
66285
3,3-diethoxyprop-1-yne
CCOC(C#C)OCC
10160-87-9
C7H12O2
128.17
MFCD00009237
RGUXEWWHSQGVRZ-UHFFFAOYSA-N
3,3-diethoxy-1-propyne,propargylaldehyde diethyl acetal,3,3-diethoxypropyne,propiolaldehyde diethyl acetal,1-propyne, 3,3-diethoxy,3,3-diethoxy-propyne,propynal diethyl acetal,1,1-diethoxyprop-2-yne,1-propyne,3,3-diethoxy,propargyl aldehyde diethylacetal
66285
3,3-diethoxyprop-1-yne
CCOC(C#C)OCC
Infrared Spectrum Authentic
Refractive Index 1.4110 to 1.4130
Density 0.8940g/mL
Specific Gravity 0.894
Boiling Point 137.0°C to 139.0°C
Packaging Glass bottle
Color Yellow
Flash Point 31°C
Quantity 1g
Assay Percent Range 97.5% min. (GC)
Linear Formula HC≡CCH(0CH2CH3)2
Formula Weight 128.17
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Propargylaldehyde diethyl acetal
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-430-4