Learn More
Propargylamine, 98%, Thermo Scientific Chemicals
Description
Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.
Solubility
Miscible with water, chloroform and ethyl acetate.
Notes
Hygroscopic. Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
Specifications
Density | 0.86 |
Boiling Point | 83°C |
Flash Point | 6°C (43°F) |
Refractive Index | 1.449 |
Linear Formula | HC≡CCH2NH2 |
Quantity | 25 g |
UN Number | UN3286 |
Beilstein | 773681 |
Sensitivity | Air Sensitive |
Solubility Information | Miscible with water,chloroform and ethyl acetate. |
Show More |
Safety and Handling
Hazard Category | H225-H302-H311-H314 |
Hazard Statement | GHS H Statement H225-H310-H314-H318-H302 Highly flammable liquid and vapor. Fatal in contact with skin. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S. |
EINECSNumber | 219-513-8 |
RTECSNumber | UK5250000 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.