Learn More
Properdin Mouse anti-Human, Clone: 3A3E1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262806
Catalog No. NBP262806
Add to cart
Description
Description
Properdin Monoclonal antibody specifically detects Properdin in Human samples. It is validated for Western Blot, Functional, Immunoassay.
Specifications
Specifications
Properdin
3A3E1
Western Blot 1:100-1:1000, Functional, Immunoassay
Purified
CFP
Human properdin purified from human plasma.
Protein G purified
Store at 4degreesC.
Monoclonal
Human
Functional Assay, In Situ Hybridization (ISH), Western Blot
Unconjugated
PBS with 0.1% BSA with No Preservative
BFD, Complement factor P, complement factor properdin, PFCcomplement, PFD, PROPERDIN
Mouse
IgG1
100μg
Primary
5199