Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propioin 95.0+%, TCI America™

Click to view available options
Quantity:
5mL
25mL
4984-85-4
C6H12O2
116.16
MFCD00043572
SKCYVGUCBRYGTE-UHFFFAOYSA-N
4-Hydroxy-3-hexanone
95609
CHEBI:18351
4-hydroxyhexan-3-one
CCC(C(=O)CC)O
4984-85-4
C6H12O2
116.16
MFCD00043572
SKCYVGUCBRYGTE-UHFFFAOYSA-N
4-Hydroxy-3-hexanone
95609
CHEBI:18351
4-hydroxyhexan-3-one
CCC(C(=O)CC)O
Color Yellow
Boiling Point 167°C
UN Number 1993
Quantity 25mL
Formula Weight 116.16
Physical Form Clear Liquid at 20°C
Percent Purity ≥95.0% (GC)
Chemical Name or Material Propioin