Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propiolic Acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10mL
25mL
100mL
Packaging:
Glass bottle
471-25-0
C3H2O2
70.04
MFCD00004360
UORVCLMRJXCDCP-UHFFFAOYSA-N
propiolic acid,2-propynoic acid,propynoic acid,propargylic acid,acetylenecarboxylic acid,carboxyacetylene,propiolic acidd,propiolicacid,unii-p2qw39g9lz,propynoate
10110
CHEBI:33199
prop-2-ynoic acid
C#CC(=O)O
471-25-0
C3H2O2
70.04
MFCD00004360
UORVCLMRJXCDCP-UHFFFAOYSA-N
propiolic acid,2-propynoic acid,propynoic acid,propargylic acid,acetylenecarboxylic acid,carboxyacetylene,propiolic acidd,propiolicacid,unii-p2qw39g9lz,propynoate
10110
CHEBI:33199
prop-2-ynoic acid
C#CC(=O)O
Flash Point 58°C
Density 1.1300g/mL
Color Colorless to Brown
Packaging Glass bottle
Melting Point 16.0°C
Quantity 25mL
Infrared Spectrum Authentic
Specific Gravity 1.13
Boiling Point 153.0°C to 157.0°C
Refractive Index 1.4315 to 1.4335
Assay Percent Range 98%
Linear Formula HC≡CCO2H
Merck Index 15, 7935
Solubility Information Solubility in water: miscible
Formula Weight 70.04
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Propiolic acid
Show More Show Less

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger